(R)-(+)-2,2'-Bis(di-2-furanylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(di-2-furylphosphine)

A115460-100MG
77245-878EA 127 USD
77245-878
(R)-(+)-2,2'-Bis(di-2-furanylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(+)-2,2'-Bis(di-2-furanylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

Caution: for Research Use Only

Formula: C₃₀H₂₄O₆P₂
MDL Number: MFCD09753004
CAS Number: 145214-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR