1-Fluoro-2-ethynylbenzene 98%

Supplier: Ambeed
Danger

Synonyms: (2-Fluorophenyl)ethyne, 1-Ethynyl-2-fluorobenzene, (2-Fluorophenyl)acetylene

A901248-10G A901248-1G A901248-25G A901248-5G
77244-714EA 38 USD
77244-714 77244-716 77244-718 77244-720
1-Fluoro-2-ethynylbenzene 98%
1-Fluoro-2-ethynylbenzene

Caution: for Research Use Only

Formula: C₈H₅F
MW: 120.13
MDL Number: MFCD00799540
CAS Number: 766-49-4
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR