1-Chlorophthalazine 95%

Supplier: Ambeed
Warning

A806675-100MG A806675-1G A806675-250MG A806675-5G
77244-242EA 47 USD
77244-242 77244-244 77244-246 77244-248
1-Chlorophthalazine 95%
1-Chlorophthalazine

Caution: for Research Use Only

Formula: C₈H₅ClN₂
MDL Number: MFCD00024141
CAS Number: 5784-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR