Bentamapimod 98%

Supplier: Ambeed
Warning

Synonyms: AS-602801, 2-(Benzo[d]thiazol-2-yl)-2-(2-((4-(morpholinomethyl)benzyl)oxy)pyrimidin-4-yl)acetonitrile

A793130-10MG A793130-1MG A793130-25MG A793130-5MG A793130-50MG
77244-160EA 63 USD
77244-160 77244-162 77244-164 77244-166 77747-545
Bentamapimod 98%
Bentamapimod

Caution: for Research Use Only

Formula: C₂₅H₂₃N₅O₂S
MDL Number: MFCD08272350
CAS Number: 848344-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR