Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane 97% (by GC)

Supplier: Ambeed
Danger

Synonyms: 1H,1H,2H,2H-Perfluorodecyltriethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane, Triethoxy-1H,1H,2H,2H-perfluorodecylsilane

A600155-1G A600155-250MG A600155-25G A600155-5G
77242-568EA 14 USD
77242-568 77242-570 77242-572 77617-516
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane 97% (by GC)
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane

Caution: for Research Use Only

Formula: C₁₆H₁₉F₁₇O₃Si
MW: 610.38 g/mol
MDL Number: MFCD00042334
CAS Number: 101947-16-4
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR