2-Chloro-5-methoxybenzimidazole 94%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methoxy-1H-benzo[d]imidazole, 2-Chloro-5-methoxy-1H-benzimidazole

A588356-250MG A588356-5G
77242-488EA 30 USD
77242-488 77242-490
2-Chloro-5-methoxybenzimidazole 94%
2-Chloro-5-methoxybenzimidazole

Caution: for Research Use Only

Formula: C₈H₇ClN₂O
MDL Number: MFCD00051945
CAS Number: 15965-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR