1-Butanesulfonyl chloride 99%

Supplier: Ambeed
Danger

Synonyms: 1-Butanesulphonylchloride, 1-Butanesulfonylchloride

A356190-25G
77240-070EA 20 USD
77240-070
1-Butanesulfonyl chloride 99%
1-Butanesulphonyl chloride

Caution: for Research Use Only

Formula: C₄H₉ClO₂S
MW: 156.63 g/mol
Boiling Pt: 208
Melting Pt: -25
MDL Number: MFCD00007464
CAS Number: 2386-60-9
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR