N-Lauroylglycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Dodecanamidoacetic acid, N-Dodecanoylglycine

A318027-5G
77239-530EA 141.11 USD
77239-530
N-Lauroylglycine 98%
N-Lauroylglycine

Caution: for Research Use Only

Formula: C₁₄H₂₇NO₃
MW: 257.37 g/mol
MDL Number: MFCD00153470
CAS Number: 7596-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR