1-Bromopyrene 97%

Supplier: Ambeed
Warning

A301466-25G A301466-5G A301466-100G A301466-10G A301466-1G
103814-304EA 5 USD
103814-304 103835-456 76838-146 76838-148 77239-246 77239-248 77305-826
1-Bromopyrene 97%
1-Bromopyrene

Caution: for Research Use Only

Formula: C₁₆H₉Br
Boiling Pt: 422.5 °C (760 mm Hg)
Melting Pt: 102…105 °C
Flash Pt: 209.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00015767
CAS Number: 1714-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR