3-Bromo-p-toluic acid 98%

Supplier: Ambeed
Danger

A231981-10G A231981-25G
77237-904EA 6 USD
77237-904 77237-906 77237-908
3-Bromo-p-toluic acid 98%
3-Bromo-p-toluic acid

Caution: for Research Use Only

Formula: C₈H₇BrO₂
MDL Number: MFCD00002488
CAS Number: 7697-26-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR