Baricitinib phosphate salt 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
LY-3009104, 2-(3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(ethylsulphonyl)azetidin-3-yl)acetonitrile phosphate, 2-(3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(ethylsulfonyl)azetidin-3-yl)acetonitrile phosphate, Baricitinib phosphate
A214136-10MG
A214136-1MG
A214136-2MG
A214136-5MG
A214136-250MG
77237-478EA
30
USD
77237-478
77237-480
77237-482
77237-484
77746-294
Baricitinib phosphate salt 98%
Baricitinib phosphate salt
Caution: for Research Use Only
Formula:
C₁₆H₁₇N₇O₂S·H₃PO₄ |
MDL Number:
MFCD25976707 CAS Number: 1187595-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...