Baricitinib phosphate salt 98%

Supplier: Ambeed
Warning

Synonyms: LY-3009104, 2-(3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(ethylsulphonyl)azetidin-3-yl)acetonitrile phosphate, 2-(3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(ethylsulfonyl)azetidin-3-yl)acetonitrile phosphate, Baricitinib phosphate

A214136-10MG A214136-1MG A214136-2MG A214136-5MG A214136-250MG
77237-478EA 30 USD
77237-478 77237-480 77237-482 77237-484 77746-294
Baricitinib phosphate salt 98%
Baricitinib phosphate salt

Caution: for Research Use Only

Formula: C₁₆H₁₇N₇O₂S·H₃PO₄
MDL Number: MFCD25976707
CAS Number: 1187595-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR