2-(4-Pyridyl)benzimidazole 98%

Supplier: Ambeed
Warning

A167748-1G A167748-25G A167748-5G
77236-172EA 13 USD
77236-172 77236-174 77236-176
2-(4-Pyridyl)benzimidazole 98%
2-(4-Pyridyl)benzimidazole

Caution: for Research Use Only

Formula: C₁₂H₉N₃
MDL Number: MFCD00453948
CAS Number: 2208-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR