1-Methylcyclopentanol 97%

Supplier: Ambeed
Danger

A144549-10G A144549-1G A144549-25G A144549-5G
103803-420EA 5 USD
103803-420 103841-496 77235-140 77299-398 77316-748 77316-750
1-Methylcyclopentanol 97%
1-Methylcyclopentanol

Caution: for Research Use Only

Formula: C₆H₁₂O
Boiling Pt: 137.1 °C (760 mm Hg)
Melting Pt: 36…37 °C
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001364
CAS Number: 1462-03-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR