(±)-1-Octen-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: (±)-Oct-1-en-3-ol, (±)-Pentyl vinyl carbinol, Oct-1-en-3-ol

A114632-100G A114632-25G A114632-500G A114632-5G A114632-10G
103824-278EA 5 USD
103824-278 77233-818 77233-820 77233-822 77311-100 77592-228
(±)-1-Octen-3-ol 98%
(±)-1-Octen-3-ol

Caution: for Research Use Only

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 174
Melting Pt: -49
Flash Pt: 61.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004589
CAS Number: 3391-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR