Baicalein 96%

Supplier: Ambeed
Warning

Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one

A106424-100G A106424-1G A106424-25G A106424-5G
77233-246EA 293 USD
77233-246 77233-248 77233-250 77233-252
Baicalein 96%
Baicalein

Caution: for Research Use Only

Formula: C₁₅H₁₀O₅
MDL Number: MFCD00017459
CAS Number: 491-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR