Anti-PEN2 Rabbit Monoclonal Antibody [clone: ARC1779]

Supplier: ANTIBODIES.COM LLC
A307565-100
77226-496EA 494.4 USD
77226-496 77226-498
Anti-PEN2 Rabbit Monoclonal Antibody [clone: ARC1779]
Antibodies
Rabbit monoclonal [ARC1779] antibody to PEN2 for WB and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2,000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: PEN2
Clonality: Monoclonal
Clone: ARC1779
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR