Anti-Pepsinogen II / PGC Rabbit Monoclonal Antibody [Clone: ARC57717]

Supplier: ANTIBODIES.COM LLC
A307491-100
77226-200EA 494.4 USD
77226-200 77226-202
Anti-Pepsinogen II / PGC Rabbit Monoclonal Antibody [Clone: ARC57717]
Antibodies
Rabbit monoclonal [ARC57717] antibody to Pepsinogen II / PGC for IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: IHC
  • Recommended Dilutions: IHC: 1:50 to 1:200

Type: Primary
Antigen: Pepsinogen II / PGC
Clonality: Monoclonal
Clone: ARC57717
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR