Anti-C9orf167 Rabbit Monoclonal Antibody [clone: ARC2939]

Supplier: ANTIBODIES.COM LLC
A307166-100
77224-916EA 494.4 USD
77224-916 77224-918
Anti-C9orf167 Rabbit Monoclonal Antibody [clone: ARC2939]
Antibodies
Rabbit monoclonal [ARC2939] antibody to C9orf167 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: C9orf167
Clonality: Monoclonal
Clone: ARC2939
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR