Human SerpinB2/PAI-2 ELISA Kit

Supplier: ANTIBODIES.COM LLC

A310173-96
77216-816EA 555.5 USD
77216-816
Human SerpinB2/PAI-2 ELISA Kit
Assays ELISAs

Human SerpinB2/PAI-2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SerpinB2/PAI-2 in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 15 to 240 ng/mL
  • Sensitivity: 7.66 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%


Human SerpinB2/PAI-2 ELISA kit (A310173) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SerpinB2/PAI-2 in serum, plasma or other biological fluids. An antibody specific for SerpinB2/PAI-2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SerpinB2/PAI-2 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-SerpinB2/PAI-2 antibody, which also binds the SerpinB2/PAI-2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SerpinB2/PAI-2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SerpinB2/PAI-2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of SerpinB2/PAI-2 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of SerpinB2/PAI-2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR