Anti-BCKDHA Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A93179-100
77207-680EA 351.7 USD
77207-680 77215-772
Anti-BCKDHA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BCKDHA for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: BCKDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR