Anti-CRISPR-Cas9 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A92824-100
77205-238EA 351.7 USD
77205-238 77207-676
Anti-CRISPR-Cas9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CRISPR-Cas9 for WB and ICC/IF with samples derived from Human.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:500-1:1, 000, ICC/IF: 1:50-1:200

Type: Primary
Antigen: CRISPR-Cas9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR