Staticide® Low-Lint Wipes, Blue
Supplier: ACL Staticide
LF99B
77200-766CS 303.86 USD
77200-766
Staticide® Low-Lint Wipes, Blue
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Exceptional all-purpose performance that is suitable for tough jobs in any workshop or manufacturing facility.
  • 55% cellulose/45% polyester hydroentangled non-woven wipe
  • Maintains strength in wet applications
  • Compatible with ISO Class 6+ (Class 1000) cleanrooms
  • Excellent choice for general industrial manufacturing use
  • 1/4 fold packaging
Blue color shows liquids easier than white wipes to assure spills are lifted.