Staticide® Checker Verification Board
Supplier: ACL Staticide
ACL307
77200-720EA 481.64 USD
77200-720
Staticide® Checker Verification Board
Resistance Testers
The ACL 307 checker board is a convenient verification unit for ensuring the ACL 306 two-point resistance tester is accurate.
  • Metal pads with 1% resistor
  • Measuring range: 10³ to 10¹° ohms
  • Measurements over 10¹° ohms will indicate ≥10¹¹
  • Made in USA
Delivery information: Includes calibration certificate.