Staticide® Two-Point Resistance Checker
Supplier: ACL Staticide
ACL306
77200-718EA 717.81 USD
77200-718
Staticide® Two-Point Resistance Checker
Resistance Testers
The Staticide ACL 306 is a convenient 2-point probe resistance tester for measuring small areas.
  • Works independent of a megohmmeter
  • Measuring range: 10³ to 10¹² ohms
  • Measures 0.375" wide
  • Made in USA
Delivery information: Includes calibration certificate.