Staticide® 8122 Premium Heel Grounder
Supplier: ACL Staticide
8122
77200-600EA 6.57 USD
77200-600
Staticide® 8122 Premium Heel Grounder
Grounders Heel and Toe Grounders
The premium heel grounder provides added conductivity, strength and longevity thanks to an extra-wide grounding strap.
  • Two-layer heel rubber
  • Wide cup-style
  • Reinforced with steel rivets
  • 1-meg ohm resistor
  • Black conductive exterior: 10⁴ ohms
This premium heel grounder features a conductive ground ribbon measuring 14.5×0.05", and can be secured with an adjustable hook and loop fabric. It is specifically designed for use by grounding employees on the move in electronic manufacturing areas.