Staticide® 8113 Premium Wrists Strap, 4 mm
Supplier: ACL Staticide
8113
77200-590EA 55.13 USD
77200-590
Staticide® 8113 Premium Wrists Strap, 4 mm
Grounders Wrist Grounders
This adjustable premium grounding wrist strap offers comfort and security for all-day use in EPAs with stringent requirements.
  • 2.5 mm band thickness for extra cushion comfort
  • Elastic crochet fabric lined with 10 rows of micro-threaded silver yarn
  • Curved buckle back for maximum comfort
  • 4 mm to banana, single wire
Designed for use with the 8107 wrist strap cord.
Packaging: 6 wrist straps per pack.