Staticide® 8111 Economy Wrist Strap, 4 mm
Supplier: ACL Staticide
8111
77200-588EA 5.36 USD
77200-588
Staticide® 8111 Economy Wrist Strap, 4 mm
Grounders Wrist Grounders
Adjustable economy ESD wrist straps offer secure personal grounding for occasional use in electrostatic protected areas and manufacturing.
  • 1.5 mm band thickness
  • Equipped with a 1-meg ohm resistor
  • 4 mm to banana, single wire
  • Use with 8101 wrist strap cord
Recommended for occasional wear, the 8111 economy wrist strap is lined with 5 rows of double-stitched silver yarn and features a flat buckle back. It is designed for use with the 8101 wrist strap cord.
Packaging: 6 wrist straps per pack.