Staticide® 8107 Premium Wrist Strap Cord, 4 mm to Banana
Supplier: ACL Staticide
8107
77200-584PK 57.18 USD
77200-584
Staticide® 8107 Premium Wrist Strap Cord, 4 mm to Banana
Grounders Grounding Wires
Heavy-duty grounding cord with 1-meg ohm resistor.
  • Heavy-duty design
  • 6 foot cord with flexible strain relief assembly to prevent breakage
  • Recommended for use with 4 mm wrist strap
  • 4 mm socket-to-banana
  • Includes alligator clip
  • Anti-static nylon
The 8107 premium wrist strap cord is desiged for use with single-wire wrist straps with 4 mm male stud.
Packaging: 6 cords per pack.