Staticide® 8102 Economy Wriststrap Cord, 10 mm to Banana
Supplier: ACL Staticide
8100
77200-576EA 38.72 USD
77200-576
Staticide® 8102 Economy Wriststrap Cord, 10 mm to Banana
Grounders Grounding Wires
Economical grounding cord with 1-meg ohm resistor.
  • 6 foot cord with flexible strain relief assembly to prevent breakage
  • Recommended for use with 10 mm wriststraps
  • 10 mm socket-to-banana
  • Includes alligator clip
  • Anti-static nylon
The 8102 Economy wriststrap cord is equipeed with a 10 mm female socket. Fits to single-wires wrist straps with 10 mm male stud.
Packaging: 6 cords per pack.