Staticide® Ground Cord for ESD Mats, 10 mm Stud Snap
Supplier: ACL Staticide
8090
77200-568EA 16.12 USD
77200-568
Staticide® Ground Cord for ESD Mats, 10 mm Stud Snap
Grounders Grounding Wires
The 8090 Mat ground cord provides a simple connection between the mat and ground source.
  • 10' coated high-strength tensile wire
  • 1-meg ohm resistor
  • 10 mm male stud connection to mats
  • Ring-style termination for connecting to grounding post or bar