Staticide® 7250 Microfiber Foam Swabs with ESD Handle
Supplier: ACL Staticide
7250
77200-558CS 332.03 USD
77200-558
Staticide® 7250 Microfiber Foam Swabs with ESD Handle
Swabs Swabs for Clinical Use
ESD-safe handle and and microfiber head for exceptionally clean critical cleaning.
  • Ultra-low particles, lint-free
  • Non-abrasive
  • ESD rigid handle, measures 10e8 -10e10 ohms
  • Class 10 cleanroom laundered
  • ISO Class 4-7 compatible
The 7250 microfiber foam swab features a small round tip constructed of medical-grade foam wrapped in microfiber knit. The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Packaging: Swabs are packaged 500 per bag, 2 bags per case.