Staticide® Polyurethane Foam Swab with ESD Handle
Supplier: ACL Staticide
7240
77200-556CS 153.19 USD
77200-556
Staticide® Polyurethane Foam Swab with ESD Handle
Swabs Swabs for Clinical Use
Medical-grade foam tipped swab with superior solvent capacity.
  • High solvent capacity
  • Non-abrasive medical grade foam
  • Blue polypropylene handle has resistance of 10⁸ to 10¹° ohms
  • Class 10 laundered and packaged
  • ISO Class 4 - 7 compatible
The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Packaging: Swabs are packaged 500 per bag, 2 bags per case.