Staticide® 7060 Polyester Swab
Supplier: ACL Staticide
7060
77200-552CS 295.68 USD
77200-552
Staticide® 7060 Polyester Swab
Swabs Swabs for Clinical Use
Robust and reliable knit swab for cleanroom applications.
  • Non-abrasive, double-layer polyester knit tip
  • Ultra-low particle; lint-free
  • Free of silicon, amides, and DOP contamination
  • White polypropylene handles
  • Class 10 laundered and packaged
  • ISO Class 5 suitable
The swab construction features ultra sonic welding, with no adhesives or binders utilized.