Staticide® 7022 Polyurethane Foam Swab
Supplier: ACL Staticide
7022
77200-550CS 557.23 USD
77200-550
Staticide® 7022 Polyurethane Foam Swab
Swabs Swabs for Clinical Use
Medical-grade foam tipped swab with superior solvent capacity.
  • High solvent capacity
  • Non-abrasive medical-grade foam
  • White polypropylene handles
  • Class 10 laundered and packaged
  • ISO Class 4-7 compatible
The swab construction features ultra sonic welding, with no adhesives or binders utilized.