1-(2-Methoxyphenyl)-2-thiourea 98%

Supplier: Ambeed
Danger

Synonyms: N-(2-Methoxyphenyl)thiourea, 1-(2-Methoxyphenyl)thiourea, 2-Methoxyphenylthiourea

A894318-1G A894318-25G A894318-5G
77200-184EA 13 USD
77200-184 77200-186 77200-188
1-(2-Methoxyphenyl)-2-thiourea 98%
1-(2-Methoxyphenyl)-2-thiourea

Caution: for Research Use Only

Formula: (C₆H₄(OCH₃))NC(S)NH₂
MDL Number: MFCD00022162
CAS Number: 1516-37-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR