Trigonelline hydrochloride 97%

Supplier: Ambeed
Warning

A764743-100MG A764743-1MG
77199-336EA 19 USD
77199-336 77199-338
Trigonelline hydrochloride 97%
Trigonelline hydrochloride

Caution: for Research Use Only

Formula: C₇H₈ClNO₂
MDL Number: MFCD00077250
CAS Number: 6138-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR