1-Allyl-2-methylbenzene 95%

Supplier: Ambeed
Danger

A506839-1G A506839-25G A506839-5G A506839-250MG
77197-078EA 29 USD
77197-078 77197-080 77197-082 77747-045
1-Allyl-2-methylbenzene 95%
1-Allyl-2-methylbenzene

Caution: for Research Use Only

Formula: C₁₀H₁₂
MDL Number: MFCD00060911
CAS Number: 1587-04-8
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR