p-Xylylenediamine 98%

Supplier: Ambeed
Danger

A466103-1G
103839-052EA 22 USD
103839-052 77196-626
p-Xylylenediamine 98%
p-Xylylenediamine

Caution: for Research Use Only

Formula: C₈H₁₂N₂
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009821
CAS Number: 539-48-0
UN: 3259
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR