trans-1,4-Cyclohexanedicarboxylic acid 97%

Supplier: Ambeed
Warning

A206067-10G A206067-25G A206067-5G A206067-100G
77192-488EA 6 USD
77192-488 77192-490 77192-492 77593-344
trans-1,4-Cyclohexanedicarboxylic acid 97%
trans-1,4-Cyclohexanedicarboxylic acid

Caution: for Research Use Only

Formula: C₈H₁₂O₄
MDL Number: MFCD00066203
CAS Number: 619-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR