R428 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(6,7-Dihydro-5H-benzo[6,7]cyclohepta[1,2-c]pyridazin-3-yl)-N3-(7-(pyrrolidin-1-yl)-6,7,8,9-tetrahydro-5H-benzo[7]annulen-2-yl)-1H-1,2,4-triazole-3,5-diamine, Bemcentinib

A171244-1MG A171244-250MG A171244-2MG A171244-50MG A171244-10MG A171244-5MG
77191-422EA 22 USD
77191-422 77191-424 77191-426 77191-428 77615-432 77746-136
R428 98%
R428

Caution: for Research Use Only

Formula: C₃₀H₃₄N₈
MDL Number: MFCD21608463
CAS Number: 1037624-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR