6-Chloro-m-toluidine 97%

Supplier: Ambeed
Danger

A134641-10G A134641-1G A134641-25G A134641-5G
77190-144EA 9 USD
77190-144 77190-146 77190-148 77190-150
6-Chloro-m-toluidine 97%
6-Chloro-m-toluidine

Caution: for Research Use Only

Formula: C₇H₈ClN
MDL Number: MFCD00007674
CAS Number: 95-81-8
UN: 2239
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR