N-Succinimidyl Acrylate 97%
Supplier: Ambeed
|
Warning
|
A107972-10G
A107972-1G
A107972-250MG
A107972-5G
A107972-25G
77188-906EA
23
USD
77188-906
77188-908
77188-910
77188-912
77549-088
N-Succinimidyl Acrylate 97%
N-Succinimidyl Acrylate
Caution: for Research Use Only
Formula:
C₇H₇NO₄ |
MDL Number:
MFCD00078261 CAS Number: 38862-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...