1,1'-Carbonyldi(1H-1,2,4-triazole) 95%

Supplier: Ambeed
Warning

Synonyms: Di(1H-1,2,4-triazol-1-yl)methanone

A105434-100G A105434-10G A105434-25G A105434-500G A105434-5G
103822-618EA 9 USD
103822-618 77188-754 77188-756 77188-758 77188-760 77309-488
1,1'-Carbonyldi(1H-1,2,4-triazole) 95%
1,1'-Carbonyldi(1H-1,2,4-triazole)

Caution: for Research Use Only

Formula: C₅H₄N₆O
MW: 164.1249 g/mol
Melting Pt: 145…150 °C
Flash Pt: 213.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043399
CAS Number: 41864-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR