AccuTec Pro Hobby Knife and Blades
Supplier: Accutec Blades
APTL-5000-0000
77172-146CS 1015.61 USD
77172-146
AccuTec Pro Hobby Knife and Blades
Knives
AccuTec Pro No.1 hobby knife and #11 hobby blades are great for detailed cutting and trimming of lightweight materials.
  • Sure-grip barrel for comfort and control
  • Four jaw chuck
  • Heavy-duty knurled collar for durability
  • Plastic safety cap protects knife when not in use
This set includes five #11 blades in a storage tube. The hobby knife is compatible with #10, #11, #16, and #17 hobby blades.
Case dimensions: 10.44×7.88×9.75"
Packaging: Each card contains one hobby knife and five #11 hobby blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.