AccuTec Pro Single Edge Blades, 2 Facet
Supplier: Accutec Blades
APBL-7037-0000
77172-142CS 411.8 USD
77172-142
AccuTec Pro Single Edge Blades, 2 Facet
Razors and Blades
AccuTec Pro single edge blades feature two-facet edges for durability. Safety blade dispenser holds 10 blades.
  • Two-facet edge for durability
  • Transparent safety dispenser with used blade vault
  • Each dispenser contains 10 blades
Case dimensions: 3.75×7.88×15.0"
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.