L-α-Aminoadipic acid 98%

Supplier: Ambeed
Warning

Synonyms: L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid

A315685-100G A315685-1G A315685-25G A315685-5G
77167-206EA 855 USD
77167-206 77167-208 77167-210 77167-212
L-α-Aminoadipic acid 98%
L-α-Aminoadipic acid

Caution: for Research Use Only

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
MDL Number: MFCD00002636
CAS Number: 1118-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR