4-(1-Methylpiperidin-4-yl)aniline 95%

Supplier: Ambeed
Warning

A269838-1G A269838-5G A269838-100MG A269838-10G A269838-250MG
103812-978EA 39 USD
103812-978 103852-884 77092-652 77092-654 77166-398 77166-400 77166-402
4-(1-Methylpiperidin-4-yl)aniline 95%
4-(1-Methylpiperidin-4-yl)aniline

Caution: for Research Use Only

Formula: C₁₂H₁₈N₂
Flash Pt: 130.079 °C
Storage Temperature: Ambient
MDL Number: MFCD06804550
CAS Number: 454482-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR