L(+)-Glutamic acid 98%

Supplier: Ambeed
Warning

A211917-1000G A211917-100G A211917-25G A211917-500G
77164-966EA 17 USD
77164-966 77164-968 77164-970 77164-972
L(+)-Glutamic acid 98%
L(+)-Glutamic acid

Caution: for Research Use Only

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number: MFCD00002634
CAS Number: 56-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR