L(+)-Glutamic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
A211917-1000G
A211917-100G
A211917-25G
A211917-500G
77164-966EA
17
USD
77164-966
77164-968
77164-970
77164-972
L(+)-Glutamic acid 98%
L(+)-Glutamic acid
Caution: for Research Use Only
Formula:
HO₂CCH₂CH₂CH(NH₂)CO₂H MW: 147.13 g/mol Boiling Pt: 325 Melting Pt: 160 |
MDL Number:
MFCD00002634 CAS Number: 56-86-0 Merck Index: 13,04482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...